Please login to your online account to display your discounted pricing
Lovibond™ Conformance Filter Sets
Glass Standards
Manufacturer: lovibond™  ORB139630
 View more versions of this product
Catalog No. 09-053-342
  • / Pack of 5 Each
Description & Specifications
Specifications
Certifications/Compliance ISO 9001: 2000 quality management system
Material Glass
For Use With Lovibond™ PFX/PFXi-880/995 colorimeters
For Use With (Application) Allow quick and simple conformance check
Quantity 5 Filters of 0.5R 1.3Y 0.1N, 1.3R 3.3Y 0.5N, 2.5R 8.6Y 0.1N, 4.0R 22Y 0.1N, 7.1R 51Y 0.5N values
Type Lovibond™ red, yellow, neutral filter set