Please login to your online account to display your discounted pricing
Lovibond™ Colorimeter: Iron
These rugged, waterproof, single parameter colorimeters are ideal for laboratory and field testing
$738.08 - $738.08
Products
Catalog Number Mfr. No. Price Quantity    
09-053-553
lovibond™
SC45075
Each for $738.08
09-053-552
tintometer inc.
SC45070
Each for $738.08
Description & Specifications
Model SC450-70 (09-053-552)
  • Range: 0.02 - 3.0mg/L
  • Method: 1,10-Phenanthroline
  • RP056-0 powder pack reagents included (100 tests)
  • Real time clock, data logging capabilities
  • Ability to export data using the IRiM
Model SC450-75 (09-053-553)
  • Range: 0.02 - 1.8mg/L
  • Method: TPTZ
  • RP055-0 powder pack reagents included (100 tests)
  • Real time clock, data logging capabilities
  • Ability to export data using the IRiM