missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lovibond™ Colorimeter: Iron
These rugged, waterproof, single parameter colorimeters are ideal for laboratory and field testing
Supplier: Lovibond™ 276056
Description
Model SC450-70 (09-053-552)
- Range: 0.02 - 3.0mg/L
- Method: 1,10-Phenanthroline
- RP056-0 powder pack reagents included (100 tests)
- Real time clock, data logging capabilities
- Ability to export data using the IRiM
- Range: 0.02 - 1.8mg/L
- Method: TPTZ
- RP055-0 powder pack reagents included (100 tests)
- Real time clock, data logging capabilities
- Ability to export data using the IRiM
Specifications
3% Full scale | |
CE | |
Backlit LCD | |
LED | |
530, 560, 580 nm | |
1.5 in. | |
Instrument, 3 x 24 mm sample cells, Iron Ferro reagent, 10 mL sample, (100 pack), batteries, tube brush, instruction manual and a carrying case | |
15.5 cm | |
0.01 A | |
1,10-Phenanthroline | |
250 g | |
7.6 cm |
Yes | |
Automatic; 16 data sets with time and date stamp | |
6.1 x 3 x 1.5 in. (15.5 x 7.6 x 3.8 cm) | |
Iron Colorimeter | |
Laboratory and field testing | |
3.8 cm | |
6.1 in. | |
0.02 to 3.0 mg/L | |
5°C to 40°C | |
0.55 lb. | |
3 in. |