missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ Colorimeter: Iron

These rugged, waterproof, single parameter colorimeters are ideal for laboratory and field testing

Manufacturer:  Lovibond™ SC45075

 View more versions of this product

Catalog No. 09-053-553


 

Specifications Description & Specifications

Specifications

Iron colorimeter
Yes
Automatic; 16 data sets with time and date stamp
LED
0.01 Abs
6.1 in.
CE
Backlit LCD
0.55 lb.
3 in.
15.5cm
1.5 in.
6.1 x 3 x 1.5 in. (15.5 x 7.6 x 3.8cm)
7.6cm
5° to 40°C
Instrument, 3 x 24mm sample cells, iron tptz reagent, 10ml sample, (100 pack), batteries, tube brush, instruction manual and a carrying case
0.02 to 1.8mg/L
250g
Laboratory and field testing
530, 560, 580nm
3% Full scale
2 Year
3.8cm
IP68