Lovibond™ Color of Varnish Discs for Lovibond™ 605 and 705 Comparator Bodies
For measuring Gardner (Varnish) color scale
Manufacturer:  Lovibond™ ORB620C40
 View more versions of this product
Catalog No. 15-398-166B
Description
Description
For Orbeco 605 and 705 Comparator Bodies (using 10mm pathlength) for measuring Gardner (Varnish) color scale.
  • Ranges 1-9 or 9-18, in steps of one unit
  • Utilizes 1933 Gardner Method
Specifications
Specifications
For measuring Gardner (Varnish) color scale
Orbeco 605 and 705 comparator bodies
Color of Varnish disc (1.0 to 9.0)
Documents