missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ Color of Varnish Discs for Lovibond™ 605 and 705 Comparator Bodies

For measuring Gardner (Varnish) color scale

$861.77 - $861.77

Specifications

For Use With Orbeco 605 and 705 comparator bodies
For Use With (Application) For measuring Gardner (Varnish) color scale
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
15-398-166B Lovibond™
ORB620C40
Color of Varnish disc (1.0 to 9.0) Each for $861.77
Specifications

Description

For Orbeco 605 and 705 Comparator Bodies (using 10mm pathlength) for measuring Gardner (Varnish) color scale.

  • Ranges 1-9 or 9-18, in steps of one unit
  • Utilizes 1933 Gardner Method

Description & Specifications

Specifications

Orbeco 605 and 705 comparator bodies
For measuring Gardner (Varnish) color scale