Please login to your online account to display your discounted pricing

Lovibond™ COD Tube Tests; MR; Mercury Free


Premeasured unit dose reagent for water quality testing

Manufacturer: lovibond™  TT20716

 View more versions of this product

Catalog No. 09-053-312

  • / Pack of 150 Each
 

Description & Specifications

Specifications

No. of Tests 150.0
For Use With (Application) Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing
Certifications/Compliance US EPA
Shelf Life 5 Years
Range 0 to 1500mg/L
Test Method Dichromate/H2SO4
For Use With (Equipment) SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments
Accessories

5 Years