Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness LR
Accurate test kits for routine water analysis
Supplier:  Lovibond™ 414260
Catalog No. 09-053-136
Includes:
  • Calibrated sample container, 100mL
  • Total Hardness VLR Tablets (250 tablets)
  • Instructions
  • Storage Case
Description
Description
Specifications
Specifications
Routine water analysis
100
VLR Total Hardness Mini Kit
100mL calibrated sample container, total hardness VLR tablets, instructions, storage case
1 to 10mg/L