Please login to your online account to display your discounted pricing
Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness HR
Accurate test kit for routine water analysis
Manufacturer: lovibond™  ORBL414450
 View more versions of this product
Catalog No. 09-053-137
  • / Each
Description & Specifications
Specifications
Range 20 to 800mg/L
For Use With (Application) Routine water analysis
Includes Dispenser bottle, 100mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case
No. of Tests 100.0
Product Type Total Hardness Mini Kit