missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness HR

Accurate test kit for routine water analysis

$151.78 - $151.78

Specifications

Product Type Total Hardness Mini Kit
For Use With (Application) Routine water analysis
No. of Tests 100.0
View More Specs

Includes:
  • Calibrated sample container, 100mL
  • Calibrated sample container, 30mL
  • Total Hardness Tablets
  • Instructions
  • Storage case


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Range Price Quantity  
Catalog Number Mfr. No. Includes Range Price Quantity  
09-053-134 Lovibond™
ORBL414240
30mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case 5 to 500mg/L Each for $151.78
09-053-137 Lovibond™
ORBL414450
Dispenser bottle, 100mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case 20 to 800mg/L Each for $151.78
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Total Hardness Mini Kit
Routine water analysis
100.0