Please login to your online account to display your discounted pricing
Lovibond™ Borosilicate Glass Colorimetry Cells
Precision fused cells
Manufacturer: lovibond™  ORB656010
 View more versions of this product
Catalog No. 09-053-352
  • / Each
Description & Specifications
Specifications
Material Borosilicate Glass
Pathlength 33mm
Type Colorimetry Cell
For Use With Model Fs, PFX195, 880, 950 and 995 instruments with heated samples (up to 100°C)