Lovibond™ Aqua Comparitor Color Discs: Phosphate
For testing water with the Aqua Comparator
$62.89 - $62.89
Specifications
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Range Price Quantity  
Catalog Number Mfr. No. Product Type Range Price Quantity  
09-053-153 Lovibond™
ORBL146250
Phosphate HR VANA Color Disc 0 to 80mg/L Each for $62.89
09-053-152 Lovibond™
ORBL146240
Phosphate Color Disc 0 to 4mg/L Each for $62.89
Specifications
Description
Economical replacement discs make it easy to test for phosphates using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™