missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ Aqua Comparitor Color Discs: pH

For testing water with the Aqua Comparator

$67.00 - $70.42

Specifications

For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Range Price Quantity  
Catalog Number Mfr. No. Product Type Range Price Quantity  
09-053-144 View Documents Lovibond™
ORBL146100
pH Color Disc 6.5 to 8.4 pH Each for $70.42
09-053-145 View Documents Lovibond™
ORBL146110
pH Bromcresol Purple Color Disc 5.2 to 6.8 pH Each for $67.00
09-053-146 View Documents Lovibond™
ORBL146120
pH Bromthymol Blue Color Disc 6 to 7.6 pH Each for $67.00
09-053-147 View Documents Lovibond™
ORBL146130
Universal pH Color Disc 4 to 10 pH Each for $67.00
Description

Description

Economical replacement discs make it easy to test for pH using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).

Specifications

Specifications

Testing water with the Aqua comparator™
Aqua comparator™