Lovibond™ Aqua Comparitor Color Discs: Iron
For testing water with the Aqua Comparator
$62.89 - $66.08
Specifications
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Range Price Quantity  
Catalog Number Mfr. No. Product Type Range Price Quantity  
09-053-150 Lovibond™
ORBL146220
Iron HR Color Disc 0.05 to 1mg/L Each for $66.08
09-053-160 Lovibond™
ORBL146320
Iron HR Color Disc 1 to 10mg/L Each for $62.89
Specifications
Description
Economical replacement discs make it easy to test for iron using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™