Please login to your online account to display your discounted pricing
Lovibond™ Aqua Comparitor Color Discs: Iron
For testing water with the Aqua Comparator
Manufacturer: lovibond™  ORBL146320
 View more versions of this product
Catalog No. 09-053-160
  • / Each
Description & Specifications
Specifications
Range 1 to 10mg/L
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
Product Type Iron HR Color Disc