Please login to your online account to display your discounted pricing
Lovibond™ Aqua Comparitor Color Discs: Copper
For testing water with the Aqua Comparator
Manufacturer: lovibond™  ORBL146440
 View more versions of this product
Catalog No. 09-053-173
  • / Each
Description & Specifications
Specifications
Range 0 to 1mg/L
For Use With (Equipment) Aqua comparator™
For Use With (Application) Testing water with the Aqua comparator™
Product Type Copper LR Color Disc