Lovibond™ Aqua Comparitor Color Discs: Chlorine
For testing water with the Aqua Comparator
Supplier: Lovibond™ 146020
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for chlorine using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0 to 4mg/L | |
Chlorine Color Disc |