Lovibond™ 711 Nesslerizer Systems
Designed for measuring unsaturated samples that are below the sensitivity of a normal, hand-held comparator system
Manufacturer:  Lovibond™ ORB711A1
 View more versions of this product
Catalog No. 15-405-311
Description
Description
  • Long path lengths: Allows for visual measurement of parameters in which very pale colors are produced such as the color of water and low ranges of chlorine
  • An integrated prism: Brings “sample” and “standard” together into one field of view
  • Simple one button press operation: Increases bulb light with constant intensity
  • Ergonomic design: Allows Nesslerizers to be used in a comfortable position
  • Guarantees correct lighting conditions 24 hours a day
Specifications
Specifications
711 Nesslerizer System
711-A1 Nessler Body, Set of 2 Nessler Tubes (250mm) and Nessler Illuminator
Documents