Please login to your online account to display your discounted pricing

Lovastatin, Tocris

HMG-CoA reductase inhibitor

$116.01 - $475.80

Chemical Identifiers

Chemical Name or Material Lovastatin
Assay Percent Range >98%
Molecular Formula C24H36O5
Formula Weight 404.54
Synonym (2S)-(1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl-2-methyl butanoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

153010

 
r & d systems
RDS153010
10mg Each for $116.01

153050

 
r & d systems
RDS153050
50mg Each for $475.80
Description & Specifications

Specifications

Formula Weight 404.54
Quantity 10mg
Chemical Name or Material Lovastatin
Assay Percent Range >98%
Molecular Formula C24H36O5
CAS 75330-75-5
Synonym (2S)-(1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl-2-methyl butanoate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature