missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lorglumide sodium salt, 98+%

Potent and specific non-peptide cholecystokinin (CCK) receptor antagonist

$255.99 - $832.74

Chemical Identifiers

CAS 97964-56-2
Molecular Formula C22H32Cl2N2O4
Molecular Weight (g/mol) 459.408
MDL Number MFCD00083183
InChI Key IEKOTSCYBBDIJC-UHFFFAOYSA-N
Synonym lorglumide, lorglumide inn, lorglumidum latin, lorglumida spanish, ccris 6857, chembl24938, d,l-4-3,4-dichlorobenzoylamino-5-dipentylamino-5-oxo-pentanoic acid, lorglumide sodium salt, lorglumida, lorglumidum
PubChem CID 3960
IUPAC Name 4-[(3,4-dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxopentanoic acid
SMILES CCCCCN(CCCCC)C(=O)C(CCC(=O)O)NC(=O)C1=CC(=C(C=C1)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61713MB View Documents Alfa Aesar™
J61713MB
25mg Each for $255.99
 
AAJ61713MC View Documents Alfa Aesar™
J61713MC
100mg Each for $832.74
 
Specifications

Chemical Identifiers

97964-56-2
459.408
IEKOTSCYBBDIJC-UHFFFAOYSA-N
3960
CCCCCN(CCCCC)C(=O)C(CCC(=O)O)NC(=O)C1=CC(=C(C=C1)Cl)Cl
C22H32Cl2N2O4
MFCD00083183
lorglumide, lorglumide inn, lorglumidum latin, lorglumida spanish, ccris 6857, chembl24938, d,l-4-3,4-dichlorobenzoylamino-5-dipentylamino-5-oxo-pentanoic acid, lorglumide sodium salt, lorglumida, lorglumidum
4-[(3,4-dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxopentanoic acid

Specifications

White
97964-56-2
MFCD00083183
IEKOTSCYBBDIJC-UHFFFAOYSA-N
4-[(3,4-dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxopentanoic acid
3960
Powder
Lorglumide sodium salt
25mg
C22H32Cl2N2O4
lorglumide, lorglumide inn, lorglumidum latin, lorglumida spanish, ccris 6857, chembl24938, d,l-4-3,4-dichlorobenzoylamino-5-dipentylamino-5-oxo-pentanoic acid, lorglumide sodium salt, lorglumida, lorglumidum
CCCCCN(CCCCC)C(=O)C(CCC(=O)O)NC(=O)C1=CC(=C(C=C1)Cl)Cl
459.408
481.39
≥98%
Safety and Handling

Safety and Handling

RTECSNumber : SA3680700