Please login to your online account to display your discounted pricing

Loreclezole hydrochloride, Tocris


Subtype-selective GABAA receptor modulator

Quantity 10mg

Manufacturer: r & d systems  129510

 View more versions of this product

Catalog No. 129510

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 311
Quantity 10mg
Chemical Name or Material Loreclezole hydrochloride
Assay Percent Range >99%
Molecular Formula C10H6Cl3N3·HCl
CAS 117857-45-1
Synonym (Z)-1-[2-Chloro-2-(2,4-dichlorophenyl)ethenyl]-1H-1,2,4-triazole hydrochloride
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Loreclezole hydrochloride
Assay Percent Range >99%
Molecular Formula C10H6Cl3N3·HCl
Formula Weight 311
Synonym (Z)-1-[2-Chloro-2-(2,4-dichlorophenyl)ethenyl]-1H-1,2,4-triazole hydrochloride
Accessories

Desiccate at -20°C