Please login to your online account to display your discounted pricing

Alfa Aesar™ Lorcainide hydrochloride, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67098MA

 View more versions of this product

Catalog No. AAJ67098MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 407.38
Assay 98%
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material Lorcainide hydrochloride
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
CAS 58934-46-6
MDL Number MFCD00055196
Color White
Quantity 10mg
Synonym N-(4-Chlorophenyl)-N-[1-(1-isopropyl)-4-piperidinyl]phenylacetamide hydrochloride
Physical Form Solid

Specifications

Chemical Name or Material Lorcainide hydrochloride
CAS 58934-46-6
Formula Weight 407.38
MDL Number MFCD00055196
Synonym N-(4-Chlorophenyl)-N-[1-(1-isopropyl)-4-piperidinyl]phenylacetamide hydrochloride
Accessories

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

SDS & Safety Documents