missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Loratadine, TCI America™

Manufacturer:  TCI America L0223100MG

 View more versions of this product

Catalog No. L0223100MG



Specifications

Specifications

Loratadine
137°C
79794-75-5
MFCD00672869
JCCNYMKQOSZNPW-UHFFFAOYSA-N
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
3957
Crystal-Powder at 20°C
White
100mg
C22H23ClN2O2
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
382.888
382.89
≥98.0% (HPLC,T)

Chemical Identifiers

79794-75-5
382.888
JCCNYMKQOSZNPW-UHFFFAOYSA-N
3957
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
C22H23ClN2O2
MFCD00672869
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
Safety and Handling

Safety and Handling

RTECSNumber : TM6129200

TSCA : No

Documents