missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Loperamide hydrochloride, 98+%

$165.01 - $615.31

Chemical Identifiers

CAS 34552-83-5
Molecular Formula C29H34Cl2N2O2
Molecular Weight (g/mol) 513.503
MDL Number MFCD00058581
InChI Key PGYPOBZJRVSMDS-UHFFFAOYSA-N
Synonym loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
PubChem CID 71420
ChEBI CHEBI:6533
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
SMILES CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6016806 View Documents Alfa Aesar™
J6016806
5g Each for $165.01
 
AAJ6016814 View Documents Alfa Aesar™
J6016814
25g Each for $615.31
 
Description

Description

  • At nanomolar concentrations, binds to mu-opioid receptors
  • L-type calcium channel blocker
Specifications

Chemical Identifiers

34552-83-5
513.503
PGYPOBZJRVSMDS-UHFFFAOYSA-N
71420
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
C29H34Cl2N2O2
MFCD00058581
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
CHEBI:6533
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl

Specifications

White
34552-83-5
MFCD00058581
14,5571
PGYPOBZJRVSMDS-UHFFFAOYSA-N
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
71420
513.51
≥98%
5g
C29H34Cl2N2O2
UN2811
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl
513.503
CHEBI:6533
Powder
Loperamide hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : TM4960000