Learn More
Loperamide hydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6016806
Description
At nanomolar concentrations, binds to mu-opioid receptors. L-type calcium channel blocker
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- At nanomolar concentrations, binds to mu-opioid receptors
- L-type calcium channel blocker
Specifications
Loperamide hydrochloride | |
White | |
MFCD00058581 | |
UN2811 | |
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec | |
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl | |
513.503 | |
CHEBI:6533 | |
≥98% |
34552-83-5 | |
C29H34Cl2N2O2 | |
5 g | |
14,5571 | |
PGYPOBZJRVSMDS-UHFFFAOYSA-N | |
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride | |
71420 | |
513.51 | |
Powder |
Chemical Identifiers
34552-83-5 | |
513.503 | |
PGYPOBZJRVSMDS-UHFFFAOYSA-N | |
71420 | |
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride |
C29H34Cl2N2O2 | |
MFCD00058581 | |
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec | |
CHEBI:6533 | |
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 252-082-4
RTECSNumber : TM4960000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only