Please login to your online account to display your discounted pricing

Lonidamine, Tocris

Mitochondrial hexokinase inhibitor

$116.01 - $116.01

Chemical Identifiers

Chemical Name or Material Lonidamine
Assay Percent Range >98%
Molecular Formula C15H10Cl2N2O2
Formula Weight 321.16
Synonym 1-[(2, 4-Dichlorophenyl)methyl]-1H-indazole-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

164610

 
r & d systems
164610
10mg Each for $116.01
Description & Specifications

Specifications

Formula Weight 321.16
Quantity 10mg
Chemical Name or Material Lonidamine
Assay Percent Range >98%
Molecular Formula C15H10Cl2N2O2
Recommended Storage Store at Room Temperature
CAS 50264-69-2
Synonym 1-[(2, 4-Dichlorophenyl)methyl]-1H-indazole-3-carboxylic acid
Safety and Handling

Store at Room Temperature