missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lomustine, TCI America™

Manufacturer:  TCI America L02511G

 View more versions of this product

Catalog No. L02511G



Specifications

Specifications

Lomustine
91°C
13010-47-4
MFCD00012392
1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea
C1CCC(CC1)NC(=O)N(CCCl)N=O
233.696
CHEBI:6520
Crystal-Powder at 20°C
Yellow
1g
C9H16ClN3O2
2811
GQYIWUVLTXOXAJ-UHFFFAOYSA-N
1-(2-chloroethyl)-3-cyclohexyl-1-nitrosourea
3950
233.70
≥98.0% (HPLC)

Chemical Identifiers

13010-47-4
233.696
GQYIWUVLTXOXAJ-UHFFFAOYSA-N
3950
1-(2-chloroethyl)-3-cyclohexyl-1-nitrosourea
C9H16ClN3O2
MFCD00012392
1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea
CHEBI:6520
C1CCC(CC1)NC(=O)N(CCCl)N=O
Safety and Handling

Safety and Handling

RTECSNumber : YS4900000

TSCA : No

Recommended Storage : Freezer

Documents