missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lomitapide, TCI America™

$250.56 - $751.69

Chemical Identifiers

CAS 182431-12-5
Molecular Formula C39H37F6N3O2
Molecular Weight (g/mol) 693.734
MDL Number MFCD16620494
InChI Key MBBCVAKAJPKAKM-UHFFFAOYSA-N
Synonym N-(2,2,2-Trifluoroethyl)-9-[4-[4-[4′-(trifluoromethyl)-[1,1′-biphenyl]-2-ylcarboxamido]piperidin-1-yl]butyl]-9H-fluorene-9-carboxamide, AEGR-733, BMS-201038
PubChem CID 9853053
ChEBI CHEBI:72297
IUPAC Name N-(2,2,2-trifluoroethyl)-9-[4-[4-[[2-[4-(trifluoromethyl)phenyl]benzoyl]amino]piperidin-1-yl]butyl]fluorene-9-carboxamide
SMILES C1CN(CCC1NC(=O)C2=CC=CC=C2C3=CC=C(C=C3)C(F)(F)F)CCCCC4(C5=CC=CC=C5C6=CC=CC=C64)C(=O)NCC(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L029825MG View Documents TCI America
L029825MG
25mg Each for $250.56
 
L0298100MG View Documents TCI America
L0298100MG
100mg Each for $751.69
 
Specifications

Chemical Identifiers

182431-12-5
693.734
MBBCVAKAJPKAKM-UHFFFAOYSA-N
9853053
N-(2,2,2-trifluoroethyl)-9-[4-[4-[[2-[4-(trifluoromethyl)phenyl]benzoyl]amino]piperidin-1-yl]butyl]fluorene-9-carboxamide
C39H37F6N3O2
MFCD16620494
N-(2,2,2-Trifluoroethyl)-9-[4-[4-[4′-(trifluoromethyl)-[1,1′-biphenyl]-2-ylcarboxamido]piperidin-1-yl]butyl]-9H-fluorene-9-carboxamide, AEGR-733, BMS-201038
CHEBI:72297
C1CN(CCC1NC(=O)C2=CC=CC=C2C3=CC=C(C=C3)C(F)(F)F)CCCCC4(C5=CC=CC=C5C6=CC=CC=C64)C(=O)NCC(F)(F)F

Specifications

White
25mg
C39H37F6N3O2
N-(2,2,2-Trifluoroethyl)-9-[4-[4-[4′-(trifluoromethyl)-[1,1′-biphenyl]-2-ylcarboxamido]piperidin-1-yl]butyl]-9H-fluorene-9-carboxamide, AEGR-733, BMS-201038
C1CN(CCC1NC(=O)C2=CC=CC=C2C3=CC=C(C=C3)C(F)(F)F)CCCCC4(C5=CC=CC=C5C6=CC=CC=C64)C(=O)NCC(F)(F)F
693.734
CHEBI:72297
Crystal-Powder at 20°C
Lomitapide
142°C
182431-12-5
MFCD16620494
MBBCVAKAJPKAKM-UHFFFAOYSA-N
N-(2,2,2-trifluoroethyl)-9-[4-[4-[[2-[4-(trifluoromethyl)phenyl]benzoyl]amino]piperidin-1-yl]butyl]fluorene-9-carboxamide
9853053
693.73
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator