missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lomefloxacin Hydrochloride, TCI America™

Manufacturer:  TCI America L02165G

 View more versions of this product

Catalog No. L02165G



Specifications

Specifications

Lomefloxacin Hydrochloride
5g
C17H20ClF2N3O3
1-Ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Hydrochloride
CCN1C=C(C(=O)C2=CC(=C(C(=C21)F)N3CCNC(C3)C)F)C(=O)O.Cl
387.812
CHEBI:6518
Crystal-Powder at 20°C
White-Yellow
98079-52-8
MFCD00214312
KXEBLAPZMOQCKO-UHFFFAOYSA-N
1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
68624
387.81
≥98.0% (HPLC,T)

Chemical Identifiers

98079-52-8
387.812
KXEBLAPZMOQCKO-UHFFFAOYSA-N
68624
1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
C17H20ClF2N3O3
MFCD00214312
1-Ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Hydrochloride
CHEBI:6518
CCN1C=C(C(=O)C2=CC(=C(C(=C21)F)N3CCNC(C3)C)F)C(=O)O.Cl
Safety and Handling

Safety and Handling

RTECSNumber : VB1997500

TSCA : No

Documents