Please login to your online account to display your discounted pricing

Lofepramine, Tocris

5-HT and noradrenalin re-uptake inhibitor (SNRI)

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Lofepramine
Assay Percent Range >99%
Molecular Formula C26H27N2OCl
Formula Weight 418.97
Synonym 1-(4-Chlorophenyl)-2-[[3-(10, 11-dihydro-5H-dibenz(Z)[b, f]azepin-5-yl)propyl]methylamino]ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

254550

 
r & d systems
254550
50mg Each for $682.67

254510

 
r & d systems
254510
10mg Each for $164.61
Description & Specifications

Specifications

Formula Weight 418.97
Quantity 50mg
Chemical Name or Material Lofepramine
Molecular Formula C26H27N2OCl
Recommended Storage Store at +4°C
Synonym 1-(4-Chlorophenyl)-2-[[3-(10, 11-dihydro-5H-dibenz(Z)[b, f]azepin-5-yl)propyl]methylamino]ethanone
CAS 23047-25-8
Assay Percent Range >99%
Safety and Handling

Store at +4°C