missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Locostatin, Thermo Scientific Chemicals
$252.21 - $763.53
Chemical Identifiers
CAS | 133812-16-5 |
---|---|
Molecular Formula | C14H15NO3 |
Molecular Weight (g/mol) | 245.278 |
MDL Number | MFCD00278769 |
InChI Key | UTZAFVPPWUIPBH-QSLRECBCSA-N |
Synonym | locostatin, s-4-benzyl-3-but-2-enoyl oxazolidin-2-one, s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one, 4s-n-crotonyl-4-benzyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone, 4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one, cell sheet migration inhibitor, locostatin, 4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one, locostatin hplc, 3-crotonyl-4beta-benzyloxazolidine-2-one |
PubChem CID | 5702600 |
IUPAC Name | (4S)-4-benzyl-3-[(E)-but-2-enoyl]-1,3-oxazolidin-2-one |
SMILES | CC=CC(=O)N1C(COC1=O)CC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64425MA
|
Thermo Scientific Chemicals
J64425MA |
10 mg |
Each for $252.21
|
|
|||||
AAJ64425MF
|
Thermo Scientific Chemicals
J64425MF |
50 mg |
Each for $763.53
|
|
|||||
Description
Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Locostatin is a cell-permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
Chemical Identifiers
133812-16-5 | |
245.278 | |
UTZAFVPPWUIPBH-QSLRECBCSA-N | |
5702600 | |
CC=CC(=O)N1C(COC1=O)CC2=CC=CC=C2 |
C14H15NO3 | |
MFCD00278769 | |
locostatin, s-4-benzyl-3-but-2-enoyl oxazolidin-2-one, s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one, 4s-n-crotonyl-4-benzyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone, 4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one, cell sheet migration inhibitor, locostatin, 4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one, locostatin hplc, 3-crotonyl-4beta-benzyloxazolidine-2-one | |
(4S)-4-benzyl-3-[(E)-but-2-enoyl]-1,3-oxazolidin-2-one |
Specifications
133812-16-5 | |
C14H15NO3 | |
10 mg | |
UTZAFVPPWUIPBH-QSLRECBCSA-N | |
(4S)-4-benzyl-3-[(E)-but-2-enoyl]-1,3-oxazolidin-2-one | |
5702600 | |
Powder |
White | |
MFCD00278769 | |
locostatin, s-4-benzyl-3-but-2-enoyl oxazolidin-2-one, s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one, 4s-n-crotonyl-4-benzyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone, 4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one, cell sheet migration inhibitor, locostatin, 4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one, locostatin hplc, 3-crotonyl-4beta-benzyloxazolidine-2-one | |
CC=CC(=O)N1C(COC1=O)CC2=CC=CC=C2 | |
245.278 | |
245.27 | |
Locostatin |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only