missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lobenzarit Disodium Salt, TCI America™

Manufacturer:  TCI America L0278100MG

 View more versions of this product

Catalog No. L0278100MG



Specifications

Specifications

Lobenzarit Disodium Salt
388°C
64808-48-6
MFCD00941422
QPJBONAWFAURGB-UHFFFAOYSA-L
disodium;2-(2-carboxylatoanilino)-4-chlorobenzoate
47462
Crystal-Powder at 20°C
White-Yellow
100mg
C14H8ClNNa2O4
N-(2-Carboxyphenyl)-4-chloroanthranilic Acid Disodium Salt, Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate
C1=CC=C(C(=C1)C(=O)[O-])NC2=C(C=CC(=C2)Cl)C(=O)[O-].[Na+].[Na+]
335.651
335.65
≥98.0% (HPLC,T)

Chemical Identifiers

64808-48-6
335.651
QPJBONAWFAURGB-UHFFFAOYSA-L
47462
C1=CC=C(C(=C1)C(=O)[O-])NC2=C(C=CC(=C2)Cl)C(=O)[O-].[Na+].[Na+]
C14H8ClNNa2O4
MFCD00941422
N-(2-Carboxyphenyl)-4-chloroanthranilic Acid Disodium Salt, Disodium N-(2-Carboxyphenyl)-4-chloroanthranilate
disodium;2-(2-carboxylatoanilino)-4-chlorobenzoate
Safety and Handling

Safety and Handling

RTECSNumber : DG4975520

TSCA : No

Documents