Please login to your online account to display your discounted pricing

Alfa Aesar™ Lobeline sulfate, 99%

Chemical Identifiers

Chemical Name or Material Lobeline sulfate
CAS 134-64-5
Molecular Formula C44H54N2O4·H2SO4
Formula Weight 773.01
MDL Number MFCD00135592
UN Number UN1544
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1732606

 
alfa aesar™
A1732606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1732603

 
alfa aesar™
A1732603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1732614

 
alfa aesar™
A1732614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 773.01
Chemical Name or Material Lobeline sulfate
Assay 99%
Molecular Formula C44H54N2O4·H2SO4
MDL Number MFCD00135592
CAS 134-64-5
UN Number UN1544
Health Hazard 1 Danger
Quantity 5g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
RTECS Number OJ8490170
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.