Please login to your online account to display your discounted pricing

LMK 235, Tocris™

Selective HDAC4/HDAC5 inhibitor

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material LMK 235
Assay Percent Range >98%
Molecular Formula C15H22N2O4
Formula Weight 294.35
Synonym N-[[6-(Hydroxyamino)-6-oxohexyl]oxy]-3, 5-dimethylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

483010

 
r & d systems
RDS483010
10mg Each for $195.49

483050

 
r & d systems
RDS483050
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 294.35
Quantity 10mg
CAS 1418033-25-6
Chemical Name or Material LMK 235
Assay Percent Range >98%
Synonym N-[[6-(Hydroxyamino)-6-oxohexyl]oxy]-3, 5-dimethylbenzamide
Molecular Formula C15H22N2O4
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C