Please login to your online account to display your discounted pricing

LM 22A4, Tocris™

Potent TrkB agonist

$231.77 - $967.24

Chemical Identifiers

Chemical Name or Material LM 22A4
Assay Percent Range >98%
Molecular Formula C15H21N3O6
Formula Weight 339.34
Synonym N,N',N''Tris(2-hydroxyethyl)-1,3,5-benzenetricarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

460750

 
r & d systems
RDS460750
50mg Each for $967.24

460710

 
r & d systems
RDS460710
10mg Each for $231.77
Description & Specifications

Specifications

Formula Weight 339.34
Quantity 50mg
Molecular Formula C15H21N3O6
CAS 37988-18-4
Recommended Storage Store at +4°C
Chemical Name or Material LM 22A4
Assay Percent Range >98%
Synonym N,N',N''Tris(2-hydroxyethyl)-1,3,5-benzenetricarboxamide
Safety and Handling

Store at +4°C