missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium zirconium oxide, 94% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 3937722
Description
Lithium zirconium oxide is used in the separation and absorption of carbon dioxide. It is used in the manufacture of dielectric bodies and insulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium zirconium oxide | |
12031-83-3 | |
100 g | |
MFCD00016182 | |
Insoluble in water. | |
[Li+].[Li+].[O-][Zr]([O-])=O | |
153.10 | |
153.1 | |
Odorless |
White to Cream | |
Powder | |
Li2O3Zr | |
lithium zirconate, dilithium dioxido oxo zirconium, 2li.o3zr, lithium zirconium oxide, dilithotab oxozirconiumbis olate, lithium zirconium oxide min, dilithium 1+ ion oxozirconiumbis olate, dilithium 1+ oxozirconiumbis olate | |
PSKLXRHTVCSWPJ-UHFFFAOYSA-N | |
dilithium;dioxido(oxo)zirconium | |
6093659 | |
≤94% |
Chemical Identifiers
12031-83-3 | |
153.10 | |
PSKLXRHTVCSWPJ-UHFFFAOYSA-N | |
6093659 | |
[Li+].[Li+].[O-][Zr]([O-])=O |
Li2O3Zr | |
MFCD00016182 | |
lithium zirconate, dilithium dioxido oxo zirconium, 2li.o3zr, lithium zirconium oxide, dilithotab oxozirconiumbis olate, lithium zirconium oxide min, dilithium 1+ ion oxozirconiumbis olate, dilithium 1+ oxozirconiumbis olate | |
dilithium;dioxido(oxo)zirconium |
Safety and Handling
EINECSNumber : 234-760-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only