Please login to your online account to display your discounted pricing

Lithium, wire, diameter 3.2mm, 99+%, ACROS Organics™

$240.48 - $240.48

Chemical Identifiers

Chemical Name or Material Lithium
Assay ≥99.0% (Li)
Assay Percent Range 99+%
Molecular Formula Li
Formula Weight 6.94
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC205600500

  SDS
thermo fisher scientific
AC205600500
50g Each for $240.48
Description & Specifications

Specifications

Assay ≥99.0% (Li)
Physical Form Wire
Formula Weight 6.94
Quantity 50g
Color Silver
Density 0.53g/mL
Fieser 01,570; 03,174; 04,286; 05,376; 10,234; 11,286; 13,157; 15,184; 17,161
Merck Index 15, 5576
Assay Percent Range 99+%
CAS 7439-93-2
Chemical Name or Material Lithium
Molecular Formula Li
Storage Information Product turns dark gray in air