missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium wire, 3.2mm (0.125 in.) dia., 99.8%, Na 0.03% max

Manufacturer:  Thermo Fisher Scientific 1077149

 View more versions of this product

Catalog No. AA1077149



Specifications

Specifications

Lithium wire
3.2mm (0.125 in.) Dia.
99.8%, Na 0.03% max.
MFCD00134051
WHXSMMKQMYFTQS-UHFFFAOYSA-N
lithium
3028194
6.941
4 x 25g
7439-93-2
Li
hsdb 549
[Li]
6.94
CHEBI:30145
Wire, annealed

Chemical Identifiers

7439-93-2
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
lithium
Li
MFCD00134051
hsdb 549
CHEBI:30145
[Li]
Documents