missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tungsten oxide, 99% min

Manufacturer:  Alfa Aesar™ 4336618

 View more versions of this product

Catalog No. AA4336618



Specifications

Specifications

Lithium tungsten oxide
Li2O4W
2li.wo4
[Li+].[Li+].[O-][W](=O)(=O)[O-]
261.716
261.73
≥99%
742°C
13568-45-1
MFCD00016185
SCKKBXOJPMZVPF-UHFFFAOYSA-N
dilithium;dioxido(dioxo)tungsten
6095624
Powder
3.71g/mL
50g

Chemical Identifiers

13568-45-1
261.716
SCKKBXOJPMZVPF-UHFFFAOYSA-N
6095624
[Li+].[Li+].[O-][W](=O)(=O)[O-]
Li2O4W
MFCD00016185
2li.wo4
dilithium;dioxido(dioxo)tungsten
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents