Learn More
Lithium tungsten oxide, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04336630
Description
Lithium tungsten oxide is employed for the preparation of ceramics with ultra-low sintering temperatures. It acts as a catalyst for oxidative coupling reactions. Furthermore, it is used in the production of high density aqueous polytungstate (metatungstate) solutions which are utilized for the separation of minerals and other solids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium tungsten oxide | |
13568-45-1 | |
250 g | |
MFCD00016185 | |
Soluble in water. | |
[Li+].[Li+].[O-][W](=O)(=O)[O-] | |
261.716 | |
261.73 | |
3.71 g/mL |
742°C | |
Powder | |
Li2O4W | |
lithium tungstate, dilithium dioxido dioxo tungsten, dilithotab tungstate, 2li.wo4, dilithium 1+ ion tungstate | |
SCKKBXOJPMZVPF-UHFFFAOYSA-N | |
dilithium;dioxido(dioxo)tungsten | |
6095624 | |
≥99% |
Chemical Identifiers
13568-45-1 | |
261.716 | |
SCKKBXOJPMZVPF-UHFFFAOYSA-N | |
6095624 | |
[Li+].[Li+].[O-][W](=O)(=O)[O-] |
Li2O4W | |
MFCD00016185 | |
lithium tungstate, dilithium dioxido dioxo tungsten, dilithotab tungstate, 2li.wo4, dilithium 1+ ion tungstate | |
dilithium;dioxido(dioxo)tungsten |
Safety and Handling
EINECSNumber : 236-978-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only