missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride, 0.5M solution in THF, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439118000

 View more versions of this product

Catalog No. AC439118000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Reduction reagent
Specifications

Specifications

Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride
product may develop some turbidity
lithium tris 3-ethyl-3-pentyl oxy aluminohydride
[Li+].CCC(CC)(CC)O[Al-](OC(CC)(CC)CC)OC(CC)(CC)CC
380.51
380.51
0.40 to 0.60M
AcroSeal™ Glass Bottle
Colorless
−17°C
79172-99-9,109-99-9
C21H46AlLiO3
LOQLYMHNMOOMHA-UHFFFAOYSA-N
lithium;tris(3-ethylpentan-3-yloxy)aluminum(1-)
16689417
Solution
0.9040g/mL
0.904
800mL

Chemical Identifiers

79172-99-9
380.51
lithium tris 3-ethyl-3-pentyl oxy aluminohydride
lithium;tris(3-ethylpentan-3-yloxy)aluminum(1-)
C21H46AlLiO3
LOQLYMHNMOOMHA-UHFFFAOYSA-N
16689417
[Li+].CCC(CC)(CC)O[Al-](OC(CC)(CC)CC)OC(CC)(CC)CC
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
May form explosive per

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

Documents