Please login to your online account to display your discounted pricing

Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride, 0.5M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride
CAS 79172-99-9
CAS 109-99-9
Molecular Formula C21H46AlLiO3
Formula Weight 380.51
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC439111000

 
thermo fisher scientific
AC439111000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439118000

 
thermo fisher scientific
AC439118000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapour. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. May form explosive peroxides. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.
Formula Weight 380.51
Physical Form Solution
Quantity 100mL
Color Colorless
Packaging AcroSeal™ Glass Bottle
Concentration 0.40 to 0.60M
Density 0.9040g/mL
Health Hazard 1 Danger
CAS 109-99-9
Molecular Formula C21H46AlLiO3
CAS 79172-99-9
Chemical Name or Material Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride
Additional Information product may develop some turbidity

Reduction reagent

Safety and Handling

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Highly flammable liquid and vapour. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. May form explosive peroxides. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.