Please login to your online account to display your discounted pricing

Lithium (trimethylsilyl)acetylide , 0.5M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC380651000

 View more versions of this product

Catalog No. AC380651000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 104.15
Quantity 100mL
Color Clear yellow to orange
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 0.4 to 0.6M
Density 0.8800g/mL
Chemical Name or Material Lithium (trimethylsilyl)acetylide
Molecular Formula C5H9LiSi
Synonym (Trimethylsilyl)ethynyl lithium
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se
MDL Number MFCD00075059
CAS 54655-07-1
Health Hazard 1 Danger
CAS 109-99-9
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides.

Specifications

Chemical Name or Material Lithium (trimethylsilyl)acetylide
CAS 54655-07-1
CAS 109-99-9
Molecular Formula C5H9LiSi
Formula Weight 104.15
MDL Number MFCD00075059
Synonym (Trimethylsilyl)ethynyl lithium
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se

GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides.

SDS & Safety Documents