missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium trifluoroacetate monohydrate, 97%

Manufacturer:  Alfa Aesar™ 8893809

 View more versions of this product

Catalog No. AA8893809



Specifications

Specifications

Lithium trifluoroacetate monohydrate
10g
Hygroscopic
C2F3LiO2
MFCD00149663
Trifluoroacetic acid lithium salt
[Li+].C(=O)(C(F)(F)F)[O-]
119.955
137.97 (119.96 Anhydrous)
2923-17-3
White
97%
LiCO2CF3·H2O
UN2811
HSFDLPWPRRSVSM-UHFFFAOYSA-M
lithium;2,2,2-trifluoroacetate
23661853
Powder

Chemical Identifiers

2923-17-3
119.955
HSFDLPWPRRSVSM-UHFFFAOYSA-M
23661853
[Li+].C(=O)(C(F)(F)F)[O-]
C2F3LiO2
MFCD00149663
Trifluoroacetic acid lithium salt
lithium;2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents