missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Trifluoro(trifluoromethyl)borate - Dimethyl Carbonate Complex, TCI America™

Manufacturer:  TCI America L02401G

 View more versions of this product

Catalog No. L02401G



Specifications

Specifications

Lithium Trifluoro(trifluoromethyl)borate - Dimethyl Carbonate Complex
1g
C4H6BF6LiO3
[Li+].[B-](C(F)(F)F)(F)(F)F.COC(=O)OC
233.829
233.83
≥98.0% (T)
White
1443685-69-5
ZGAWBZORCLPFJC-UHFFFAOYSA-N
lithium;dimethyl carbonate;trifluoro(trifluoromethyl)boranuide
56973698
Crystal-Powder at 20°C

Chemical Identifiers

1443685-69-5
233.829
56973698
[Li+].[B-](C(F)(F)F)(F)(F)F.COC(=O)OC
C4H6BF6LiO3
ZGAWBZORCLPFJC-UHFFFAOYSA-N
lithium;dimethyl carbonate;trifluoro(trifluoromethyl)boranuide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents