missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium triethylborohydride, 1.7M solution in THF, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450691000
Specifications
Lithium triethylborohydride | |
C6H16BLi | |
0.8900g/mL | |
Solubility in water: reacts. | |
100mL |
22560-16-3 | |
LiB(C2H5)3H | |
105.94 | |
0.89 | |
−17°C |
Chemical Identifiers
22560-16-3 | |
C6H16BLi |