missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium triethylborohydride, 1.7M solution in THF, ACROS Organics™

Chemical Identifiers

CAS 22560-16-3
Molecular Formula C6H16BLi
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC450691000
SDS View Documents
Thermo Fisher Scientific
AC450691000
100mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC450698000
SDS View Documents
Thermo Fisher Scientific
AC450698000
800mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

22560-16-3
C6H16BLi

Specifications

22560-16-3
LiB(C2H5)3H
105.94
0.89
−17°C
C6H16BLi
0.8900g/mL
Solubility in water: reacts.
100mL
Lithium triethylborohydride
SDS