missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium triethylborohydride, 1M in THF

As reducing agent

Manufacturer:  Alfa Aesar™ 89144AC

 View more versions of this product

Catalog No. AA89144AC



Specifications

Specifications

Colorless
Lithium triethylborohydride
−17°C (1°F)
22560-16-3
LiB(CH2CH3)3H
UN3399
14,5544
1M in THF
Liquid
25mL
0.892g/mL
Moisture Sensitive
C6H16BLi
MFCD00011703
4151240
Lithium triethylhydridoborate
105.94

Chemical Identifiers

22560-16-3
C6H16BLi
MFCD00011703
Lithium triethylhydridoborate
Safety and Handling

Safety and Handling

GHS H Statement
H225-H260-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

SDS Documents