Please login to your online account to display your discounted pricing

Lithium triethylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  176991000

 View more versions of this product

Catalog No. AC176991000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Cloudy Solution
Color Colorless to light gray
Formula Weight 105.94
Quantity 100mL
Merck Index 15, 5600
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 0.9 to 1.1M
EINECS Number 245-076-8
Density 0.8920g/mL
Fieser 04,313; 06,348; 08,309; 09,286; 10,249; 11,304; 12,289; 17,172
CAS 109-99-9
Synonym Lithium triethylhydroborate
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of
Health Hazard 2 GHS P Statement Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage.
Health Hazard 1 Danger
Name Note 1M Solution in THF
Chemical Name or Material Lithium triethylborohydride
MDL Number MFCD00011703
Linear Formula LiB(C2H5)3H
CAS 22560-16-3
Molecular Formula C6H16BLi

Specifications

Chemical Name or Material Lithium triethylborohydride
Name Note 1M Solution in THF
CAS 22560-16-3
CAS 109-99-9
Molecular Formula C6H16BLi
Linear Formula LiB(C2H5)3H
Formula Weight 105.94
MDL Number MFCD00011703
Synonym Lithium triethylhydroborate
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of

GHS P Statement Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage.

SDS & Safety Documents