Please login to your online account to display your discounted pricing

Lithium triethylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

$549.68 - $549.68

Chemical Identifiers

Chemical Name or Material Lithium triethylborohydride
Name Note 1M Solution in THF
CAS 22560-16-3
CAS 109-99-9
Molecular Formula C6H16BLi
Linear Formula LiB(C2H5)3H
Formula Weight 105.94
MDL Number MFCD00011703
Synonym Lithium triethylhydroborate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176998000

  SDS
thermo fisher scientific
176998000
AcroSeal™ Glass Bottle 800mL Each for $549.68

AC176991000

  SDS
thermo fisher scientific
176991000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Colorless to light gray
Physical Form Cloudy Solution
Formula Weight 105.94
Quantity 800mL
Packaging AcroSeal™ Glass Bottle
EINECS Number 245-076-8
Concentration 0.9 to 1.1M
Density 0.8920g/mL
Fieser 04,313; 06,348; 08,309; 09,286; 10,249; 11,304; 12,289; 17,172
Merck Index 15, 5600
Chemical Name or Material Lithium triethylborohydride
Name Note 1M Solution in THF
Molecular Formula C6H16BLi
Linear Formula LiB(C2H5)3H
MDL Number MFCD00011703
CAS 22560-16-3
Synonym Lithium triethylhydroborate
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage.
CAS 109-99-9
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of

Reduction reagent

Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of

GHS P Statement Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage.